Product Details

SNP ID
rs142059730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17338570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAGGGCTTCGACCGGCGGAGGC[A/G]GGCGAGTTCACCAGACGGGCGTTCG
Phenotype
MIM: 610216 MIM: 608536
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO8 PubMed Links

Gene Details

Gene
ANO8
Gene Name
anoctamin 8
There are no transcripts associated with this gene.

Gene
GTPBP3
Gene Name
GTP binding protein 3 (mitochondrial)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128855.2 515 Silent Mutation GCA,GCG A140A NP_001122327.1
NM_001195422.1 515 Silent Mutation GCA,GCG A162A NP_001182351.1
NM_032620.3 515 Silent Mutation GCA,GCG A140A NP_116009.2
NM_133644.3 515 Silent Mutation GCA,GCG A140A NP_598399.2

View Full Product Details