Product Details

SNP ID
rs142123915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16680018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGTCCCGGCGCCACCCCATCGC[A/G]TCCTGGCTGTGCGCCATGCTGCATT
Phenotype
MIM: 611235
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM38A PubMed Links

Gene Details

Gene
TMEM38A
Gene Name
transmembrane protein 38A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024074.2 250 Silent Mutation GCA,GCG A53A NP_076979.1

View Full Product Details