Product Details

SNP ID
rs142390134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18920686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATCCCTATGGCATCTTCTGCCT[C/T]GTCCTGACACCCACCAGGTAAGCCC
Phenotype
MIM: 606942 MIM: 604800
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPE PubMed Links

Gene Details

Gene
COPE
Gene Name
coatomer protein complex subunit epsilon
There are no transcripts associated with this gene.

Gene
DDX49
Gene Name
DEAD-box helicase 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019070.4 289 Silent Mutation CTC,CTT L74L NP_061943.2
XM_011528083.1 289 UTR 5 XP_011526385.1
XM_011528084.2 289 UTR 5 XP_011526386.1
Gene
HOMER3
Gene Name
homer scaffolding protein 3
There are no transcripts associated with this gene.

View Full Product Details