Product Details
- SNP ID
-
rs142391144
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:5914587 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTTTTCCGCCAACTAGACCGGGA[C/T]GGGAGCAGATCCCTGGACGCTGATG
- Phenotype
-
MIM: 114212
MIM: 603327
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CAPS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7249419] are located under a probe and SNP(s) [rs7248090] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAPS
- Gene Name
- calcyphosine
- Gene
- RANBP3
- Gene Name
- RAN binding protein 3
There are no transcripts associated with this gene.
- Gene
- VMAC
- Gene Name
- vimentin-type intermediate filament associated coiled-coil protein
There are no transcripts associated with this gene.
View Full Product Details