Product Details

SNP ID
rs142407410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45997249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGTCTGTGTGGCTTCTTCCAC[A/C]CCTCTTTCAGGTCTCCGCTCAGACG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC61 PubMed Links

Gene Details

Gene
CCDC61
Gene Name
coiled-coil domain containing 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267723.1 Intron NP_001254652.1
XM_005259191.3 Intron XP_005259248.1
XM_005259192.4 Intron XP_005259249.1
XM_011527257.2 Intron XP_011525559.1

View Full Product Details