Product Details

SNP ID
rs142423316
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6413494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGACAACAGACCAGTCCTGGGAG[A/G]GGGGACGGGCGGGGCCGCGGGAGCT
Phenotype
MIM: 603445
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KHSRP PubMed Links

Gene Details

Gene
KHSRP
Gene Name
KH-type splicing regulatory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003685.2 3774 UTR 3 NP_003676.2
XM_005259668.4 3774 UTR 3 XP_005259725.1
XM_011528395.2 3774 Intron XP_011526697.1
XM_017027408.1 3774 UTR 3 XP_016882897.1
XM_017027409.1 3774 Intron XP_016882898.1
XM_017027410.1 3774 Intron XP_016882899.1
Gene
LOC390877
Gene Name
adenylate kinase isoenzyme 1-like
There are no transcripts associated with this gene.

Gene
MIR3940
Gene Name
microRNA 3940
There are no transcripts associated with this gene.

View Full Product Details