Product Details

SNP ID
rs142492636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2790470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATCTCCGTGCTCTGTGGTAAA[C/T]GACCTGCGGTGGGACCTGAGTGCCC
Phenotype
MIM: 603419 MIM: 601117
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGTA PubMed Links

Gene Details

Gene
SGTA
Gene Name
small glutamine rich tetratricopeptide repeat containing alpha
There are no transcripts associated with this gene.

Gene
THOP1
Gene Name
thimet oligopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003249.4 269 Silent Mutation AAC,AAT N22N NP_003240.1
XM_011528228.2 269 Intron XP_011526530.1

View Full Product Details