Product Details

SNP ID
rs142578266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3808971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTGCTCACAGCCTTCTCCACCA[G/T]CAGCTCCATGGCCTGGTGGGGACAG
Phenotype
MIM: 600038
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MATK PubMed Links

Gene Details

Gene
MATK
Gene Name
megakaryocyte-associated tyrosine kinase
There are no transcripts associated with this gene.

Gene
ZFR2
Gene Name
zinc finger RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145640.1 2460 Intron NP_001139112.1
NM_015174.1 2460 Missense Mutation ATG,CTG M816L NP_055989.1
XM_011527831.2 2460 Missense Mutation ATG,CTG M817L XP_011526133.1

View Full Product Details