Product Details

SNP ID
rs142645416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35778541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCTCCACCGGTGCATATTTGA[C/T]CGGAGGTTCTCCTGCCTTCCGGAGC
Phenotype
MIM: 614902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP33 PubMed Links

Gene Details

Gene
ARHGAP33
Gene Name
Rho GTPase activating protein 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172630.1 1888 UTR 5 NP_001166101.1
NM_052948.3 1888 Silent Mutation GAC,GAT D116D NP_443180.2
XM_005258488.1 1888 Silent Mutation GAC,GAT D116D XP_005258545.1
XM_006722999.1 1888 Silent Mutation GAC,GAT D116D XP_006723062.1
XM_006723003.2 1888 Intron XP_006723066.1
XM_011526416.1 1888 UTR 5 XP_011524718.1
XM_011526417.1 1888 UTR 5 XP_011524719.1
XM_011526418.1 1888 UTR 5 XP_011524720.1
XM_011526419.1 1888 UTR 5 XP_011524721.1
XM_011526420.1 1888 UTR 5 XP_011524722.1
XM_011526421.2 1888 Silent Mutation GAC,GAT D153D XP_011524723.1
XM_017026238.1 1888 Silent Mutation GAC,GAT D153D XP_016881727.1
Gene
PROSER3
Gene Name
proline and serine rich 3
There are no transcripts associated with this gene.

View Full Product Details