Product Details

SNP ID
rs142725492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38838959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGGTAATGGCCCGGTCTACAGC[A/G]TAGCCATCAGCCATCTCCACCATGG
Phenotype
MIM: 600696 MIM: 603083
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ECH1 PubMed Links

Gene Details

Gene
ECH1
Gene Name
enoyl-CoA hydratase 1
There are no transcripts associated with this gene.

Gene
HNRNPL
Gene Name
heterogeneous nuclear ribonucleoprotein L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005335.1 Intron NP_001005335.1
NM_001533.2 Intron NP_001524.2
XM_011526886.1 Intron XP_011525188.1
XM_011526887.1 Intron XP_011525189.1
XM_011526888.2 Intron XP_011525190.1
XM_011526889.1 Intron XP_011525191.1

View Full Product Details