Product Details

SNP ID
rs142869656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44262290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTACGCTGAAGGCTTCCAAGAG[A/G]AGGGATTACAAGAGAAGGACAGGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF233 PubMed Links

Gene Details

Gene
ZNF233
Gene Name
zinc finger protein 233
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207005.1 Intron NP_001193934.1
NM_181756.2 Intron NP_861421.2
XM_017026758.1 Intron XP_016882247.1
XM_017026759.1 Intron XP_016882248.1
XM_017026760.1 Intron XP_016882249.1
XM_017026761.1 Intron XP_016882250.1

View Full Product Details