Product Details

SNP ID
rs142870685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35550245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTACCATCGCCCAGGACACAAGC[A/G]TGTCTTTAACGAAGGGCCCTCAGGC
Phenotype
MIM: 137216 MIM: 609169 MIM: 613585
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP4A PubMed Links

Gene Details

Gene
ATP4A
Gene Name
ATPase H+/K+ transporting alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000704.2 3507 UTR 3 NP_000695.2
Gene
GAPDHS
Gene Name
glyceraldehyde-3-phosphate dehydrogenase, spermatogenic
There are no transcripts associated with this gene.

Gene
TMEM147
Gene Name
transmembrane protein 147
There are no transcripts associated with this gene.

Gene
TMEM147-AS1
Gene Name
TMEM147 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details