Product Details

SNP ID
rs142878848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7076446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGAGTGGGCATTGCTGGACCC[G/T]GCCCAAAGGACACTGTACAGGGACG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF557 PubMed Links

Gene Details

Gene
ZNF557
Gene Name
zinc finger protein 557
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044387.1 671 Silent Mutation CCG,CCT P62P NP_001037852.1
NM_001044388.1 671 Silent Mutation CCG,CCT P55P NP_001037853.1
NM_024341.2 671 Silent Mutation CCG,CCT P62P NP_077317.2

View Full Product Details