Product Details

SNP ID
rs142921732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36447722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACAAAAAGAGTATTAGTCGGAAT[A/G]AAGTCCTTCACACACATTATTCACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF566 PubMed Links

Gene Details

Gene
ZNF566
Gene Name
zinc finger protein 566
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145343.1 2861 UTR 3 NP_001138815.1
NM_001145344.1 2861 UTR 3 NP_001138816.1
NM_001145345.1 2861 UTR 3 NP_001138817.1
NM_001300970.1 2861 UTR 3 NP_001287899.1
NM_032838.4 2861 UTR 3 NP_116227.1
XM_005259354.2 2861 UTR 3 XP_005259411.1
XM_005259356.2 2861 UTR 3 XP_005259413.1
XM_006723447.2 2861 UTR 3 XP_006723510.1
XM_011527428.1 2861 UTR 3 XP_011525730.1
XM_017027399.1 2861 UTR 3 XP_016882888.1
XM_017027400.1 2861 UTR 3 XP_016882889.1

View Full Product Details