Product Details

SNP ID
rs143111515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12922751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGTCCTAGAGTGGCCCATGTCA[C/T]GCAGCCTCCTGTGTGGGAGGGGGCC
Phenotype
MIM: 602918 MIM: 608801 MIM: 611487
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FARSA PubMed Links

Gene Details

Gene
FARSA
Gene Name
phenylalanyl-tRNA synthetase alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004461.2 1572 Silent Mutation GCA,GCG A508A NP_004452.1
Gene
GCDH
Gene Name
glutaryl-CoA dehydrogenase
There are no transcripts associated with this gene.

Gene
MIR5695
Gene Name
microRNA 5695
There are no transcripts associated with this gene.

Gene
SYCE2
Gene Name
synaptonemal complex central element protein 2
There are no transcripts associated with this gene.

View Full Product Details