Product Details
- SNP ID
-
rs143156732
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:8302539 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCAGTAACCCCAGTGGGCGGAGG[C/T]GCTCCTGGGCTCGGAGCCAGGACAA
- Phenotype
-
MIM: 606475
MIM: 602139
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CD320
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs146190802] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD320
- Gene Name
- CD320 molecule
- Gene
- NDUFA7
- Gene Name
- NADH:ubiquinone oxidoreductase subunit A7
There are no transcripts associated with this gene.
View Full Product Details