Product Details

SNP ID
rs143285338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6952936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAAGAAGAAAATCCACTTCTCC[C/T]GCCCGCCATCTTTAAAATGCATCCT
Phenotype
MIM: 612305
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRE4P PubMed Links

Gene Details

Gene
ADGRE4P
Gene Name
adhesion G protein-coupled receptor E4, pseudogene
There are no transcripts associated with this gene.

View Full Product Details