Product Details

SNP ID
rs143297439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45071636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCCGCCTCAGGGTGCTTCTGCC[A/G]CAGGTGTTTGTCCAGGGTGGCTCGC
Phenotype
MIM: 607419 MIM: 613226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLASRP PubMed Links

Gene Details

Gene
CLASRP
Gene Name
CLK4 associating serine/arginine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278439.1 Intron NP_001265368.1
NM_007056.2 Intron NP_008987.2
XM_011526396.2 Intron XP_011524698.1
XM_011526397.2 Intron XP_011524699.1
Gene
GEMIN7
Gene Name
gem nuclear organelle associated protein 7
There are no transcripts associated with this gene.

Gene
ZNF296
Gene Name
zinc finger protein 296
There are no transcripts associated with this gene.

View Full Product Details