Product Details

SNP ID
rs143325605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54096928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCCGGCCCCCAGTCTGGCCTTC[C/G]GTAGCAGCAGCGGTAACTTCCCCCT
Phenotype
MIM: 603832 MIM: 606862
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NDUFA3 PubMed Links

Gene Details

Gene
NDUFA3
Gene Name
NADH:ubiquinone oxidoreductase subunit A3
There are no transcripts associated with this gene.

Gene
OSCAR
Gene Name
osteoclast associated, immunoglobulin-like receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282349.1 511 Missense Mutation CGA,GGA R103G NP_001269278.1
NM_001282350.1 511 Missense Mutation CGA,GGA R92G NP_001269279.1
NM_130771.4 511 Missense Mutation CGA,GGA R107G NP_570127.3
NM_133168.4 511 Missense Mutation CGA,GGA R92G NP_573398.2
NM_133169.4 511 Missense Mutation CGA,GGA R103G NP_573399.2
NM_206818.2 511 Missense Mutation CGA,GGA R107G NP_996554.2

View Full Product Details