Product Details

SNP ID
rs143404539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43184864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGTGTAAGGTGTAGGATCCTGC[A/G]TCTTCCCGGGTGACATTCTGGATCA
Phenotype
MIM: 176393 MIM: 176394
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSG4 PubMed Links

Gene Details

Gene
PSG4
Gene Name
pregnancy specific beta-1-glycoprotein 4
There are no transcripts associated with this gene.

Gene
PSG5
Gene Name
pregnancy specific beta-1-glycoprotein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130014.1 477 Intron NP_001123486.1
NM_002781.3 477 Intron NP_002772.3
XM_011527132.2 477 Silent Mutation XP_011525434.1
XM_017027003.1 477 Intron XP_016882492.1

View Full Product Details