Product Details

SNP ID
rs143409655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2039658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCACTGGGGCCGAGGACAGACT[A/G]GCCCTTTGCCGCCGCTGCGCCAGCT
Phenotype
MIM: 605069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MKNK2 PubMed Links

Gene Details

Gene
MKNK2
Gene Name
MAP kinase interacting serine/threonine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017572.3 1598 Intron NP_060042.2
NM_199054.2 1598 Silent Mutation GCC,GCT A451A NP_951009.1

View Full Product Details