Product Details

SNP ID
rs143608128
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57507396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCTCCAGTCTTGTTAAACATC[A/G]AAGGATTCACACTGGAGAAATACAA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF773 PubMed Links

Gene Details

Gene
ZNF773
Gene Name
zinc finger protein 773
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304334.1 1525 Missense Mutation CAA,CGA Q433R NP_001291263.1
NM_001304335.1 1525 Intron NP_001291264.1
NM_001304336.1 1525 UTR 3 NP_001291265.1
NM_001304337.1 1525 Intron NP_001291266.1
NM_198542.2 1525 Missense Mutation CAA,CGA Q434R NP_940944.1

View Full Product Details