Product Details

SNP ID
rs143644123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14799331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCACTGAGGCCACCAGACTTGTC[C/T]TAGAAGATGGTGTGGCTGCAGAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR7C1 PubMed Links

Gene Details

Gene
OR7C1
Gene Name
olfactory receptor family 7 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198944.1 1310 Intron NP_945182.1
XM_005259853.3 1310 Missense Mutation AAG,AGG K269R XP_005259910.1
XM_011527915.2 1310 Missense Mutation AAG,AGG K319R XP_011526217.1

View Full Product Details