Product Details

SNP ID
rs143650831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43827433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCGGTGTCCTTAAGGTTCTT[A/G]GACTCTGCATGGTGAGTTGGTCGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYPD5 PubMed Links

Gene Details

Gene
LYPD5
Gene Name
LY6/PLAUR domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031749.2 142 Intron NP_001026919.2
NM_001288763.1 142 Intron NP_001275692.1
NM_182573.2 142 Intron NP_872379.2
XM_011526794.2 142 Intron XP_011525096.1
Gene
ZNF283
Gene Name
zinc finger protein 283
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297752.1 142 UTR 5 NP_001284681.1
NM_181845.1 142 UTR 5 NP_862828.1
XM_005258784.3 142 UTR 5 XP_005258841.1
XM_005258785.3 142 UTR 5 XP_005258842.1
XM_005258786.3 142 UTR 5 XP_005258843.1
XM_017026628.1 142 UTR 5 XP_016882117.1
XM_017026629.1 142 UTR 5 XP_016882118.1
XM_017026630.1 142 UTR 5 XP_016882119.1
XM_017026631.1 142 UTR 5 XP_016882120.1
XM_017026632.1 142 UTR 5 XP_016882121.1
XM_017026633.1 142 UTR 5 XP_016882122.1
XM_017026634.1 142 UTR 5 XP_016882123.1
XM_017026635.1 142 UTR 5 XP_016882124.1
XM_017026636.1 142 Intron XP_016882125.1
XM_017026637.1 142 UTR 5 XP_016882126.1
XM_017026638.1 142 UTR 5 XP_016882127.1
XM_017026639.1 142 UTR 5 XP_016882128.1

View Full Product Details