Product Details

SNP ID
rs143676147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49878500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGCCTCCCTCCTTACCCTAGGA[C/T]AATGACGTCCTCCTGCACTGGGCTC
Phenotype
MIM: 610221 MIM: 616659
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKT1S1 PubMed Links

Gene Details

Gene
AKT1S1
Gene Name
AKT1 substrate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098632.2 323 Intron NP_001092102.1
NM_001098633.3 323 Intron NP_001092103.1
NM_001278159.1 323 Intron NP_001265088.1
NM_001278160.1 323 Intron NP_001265089.1
NM_032375.5 323 Intron NP_115751.3
Gene
MIR4750
Gene Name
microRNA 4750
There are no transcripts associated with this gene.

Gene
TBC1D17
Gene Name
TBC1 domain family member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168222.1 323 Silent Mutation GAC,GAT D8D NP_001161694.1
NM_024682.2 323 Silent Mutation GAC,GAT D41D NP_078958.2
XM_011527317.2 323 Silent Mutation GAC,GAT D41D XP_011525619.1

View Full Product Details