Product Details

SNP ID
rs143728697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41093689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACCTGGTGATGACCACCCTGAA[C/G]CTCTTCTTTGCGGGCACTGAGACCG
Phenotype
MIM: 608055
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYP2A13 PubMed Links

Gene Details

Gene
CYP2A13
Gene Name
cytochrome P450 family 2 subfamily A member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000766.4 Intron NP_000757.2

View Full Product Details