Product Details
- SNP ID
-
rs143763081
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:11325546 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGCGCTCGAAGACCTGCTTCACA[C/T]TGATGTTCTCCTTGGCACTGGCTTC
- Phenotype
-
MIM: 604350
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RAB3D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12981706] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RAB3D
- Gene Name
- RAB3D, member RAS oncogene family
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004283.3 |
773 |
Missense Mutation |
AAT,AGT |
N171S |
NP_004274.1 |
- Gene
- TSPAN16
- Gene Name
- tetraspanin 16
View Full Product Details