Product Details

SNP ID
rs143844750
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11298214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCCTCTCTGACGAATGTCCTC[A/G]GGCTGTCCTCCGCATACCTCCTTCA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HSPC102 PubMed Links

Gene Details

Gene
HSPC102
Gene Name
uncharacterized HSPC102
There are no transcripts associated with this gene.

Gene
TSPAN16
Gene Name
tetraspanin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282509.1 232 Missense Mutation AGG,GGG R48G NP_001269438.1
NM_001282510.1 232 Missense Mutation AGG,GGG R48G NP_001269439.1
NM_012466.3 232 Missense Mutation AGG,GGG R48G NP_036598.1
XM_011527901.1 232 Missense Mutation AGG,GGG R48G XP_011526203.1
XM_011527902.1 232 Missense Mutation AGG,GGG R48G XP_011526204.1
XM_011527903.2 232 UTR 5 XP_011526205.1

View Full Product Details