Product Details

SNP ID
rs143940810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36006825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGTCTCGGTAAGCTCCCAGGGC[C/T]CGCAGGATCTGCTCCAGGGCTGCCC
Phenotype
MIM: 613304 MIM: 607382 MIM: 615535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALKBH6 PubMed Links

Gene Details

Gene
ALKBH6
Gene Name
alkB homolog 6
There are no transcripts associated with this gene.

Gene
CLIP3
Gene Name
CAP-Gly domain containing linker protein 3
There are no transcripts associated with this gene.

Gene
LOC101927572
Gene Name
uncharacterized LOC101927572
There are no transcripts associated with this gene.

Gene
SYNE4
Gene Name
spectrin repeat containing nuclear envelope family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039876.2 655 Silent Mutation CGA,CGG R181R NP_001034965.1
NM_001297735.2 655 Intron NP_001284664.1

View Full Product Details