Product Details

SNP ID
rs143983881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51146740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGTTGGATCACCAAAACAATTC[C/T]AATCCATCCTCAGGCAAAATGAGGC
Phenotype
MIM: 604410 MIM: 605640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928517 PubMed Links

Gene Details

Gene
LOC101928517
Gene Name
uncharacterized LOC101928517
There are no transcripts associated with this gene.

Gene
SIGLEC7
Gene Name
sialic acid binding Ig like lectin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277201.1 Intron NP_001264130.1
NM_014385.3 Intron NP_055200.1
NM_016543.3 Intron NP_057627.2
XM_011526721.2 Intron XP_011525023.1
Gene
SIGLEC9
Gene Name
sialic acid binding Ig like lectin 9
There are no transcripts associated with this gene.

View Full Product Details