Product Details

SNP ID
rs144118003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55341978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAGCACGAGAACTGTGCGAGAA[C/T]GACGACCTGGCCACCAGCCTCGTCC
Phenotype
MIM: 613198
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX6B2 PubMed Links

Gene Details

Gene
COX6B2
Gene Name
cytochrome c oxidase subunit 6B2
There are no transcripts associated with this gene.

Gene
KMT5C
Gene Name
lysine methyltransferase 5C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032701.3 290 Silent Mutation AAC,AAT N14N NP_116090.2
XM_005259338.3 290 UTR 5 XP_005259395.1
XM_006723442.3 290 Intron XP_006723505.1
XM_011527415.2 290 Silent Mutation AAC,AAT N14N XP_011525717.1
XM_017027388.1 290 UTR 5 XP_016882877.1
Gene
TMEM150B
Gene Name
transmembrane protein 150B
There are no transcripts associated with this gene.

View Full Product Details