Product Details

SNP ID
rs144178912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51372053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCGCTCGCTGGTGTACACCGCC[A/G]TGGCCACCACCATGGAGATGGCTGG
Phenotype
MIM: 130410 MIM: 154045 MIM: 606008
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLDND2 PubMed Links

Gene Details

Gene
CLDND2
Gene Name
claudin domain containing 2
There are no transcripts associated with this gene.

Gene
ETFB
Gene Name
electron transfer flavoprotein beta subunit
There are no transcripts associated with this gene.

Gene
LIM2
Gene Name
lens intrinsic membrane protein 2
There are no transcripts associated with this gene.

Gene
NKG7
Gene Name
natural killer cell granule protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005601.3 332 Missense Mutation ACG,ATG T109M NP_005592.1
XM_005258955.3 332 Missense Mutation ACG,ATG T74M XP_005259012.1
XM_006723228.3 332 Missense Mutation ACG,ATG T60M XP_006723291.1

View Full Product Details