Product Details

SNP ID
rs144263364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48333235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTCTACTCAGGAGATGTTACTG[A/C]TGGGCGTGGCTTGCGGGGGCTCGGA
Phenotype
MIM: 602335
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EMP3 PubMed Links

Gene Details

Gene
EMP3
Gene Name
epithelial membrane protein 3
There are no transcripts associated with this gene.

Gene
TMEM143
Gene Name
transmembrane protein 143
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303538.1 1576 Missense Mutation AGC,ATC S420I NP_001290467.1
NM_001303539.1 1576 Missense Mutation AGC,ATC S390I NP_001290468.1
NM_001303540.1 1576 Missense Mutation AGC,ATC S355I NP_001290469.1
NM_018273.3 1576 Missense Mutation AGC,ATC S455I NP_060743.2

View Full Product Details