Product Details

SNP ID
rs144482840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39025522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTTCGAAAGACACAAAGCGGAC[A/G]CCCATCTTGATGTTGGAGAACACGT
Phenotype
MIM: 609099
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO27 PubMed Links

Gene Details

Gene
FBXO27
Gene Name
F-box protein 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178820.4 861 Silent Mutation GGC,GGT G247G NP_849142.1
XM_017026290.1 861 Intron XP_016881779.1

View Full Product Details