Product Details

SNP ID
rs144502127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44327505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGTCCTTACCATAGTCTTCGCT[G/T]TTATAGAATTTATCACTACTATGGA
Phenotype
MIM: 603994
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF112 PubMed Links

Gene Details

Gene
ZNF112
Gene Name
zinc finger protein 112
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083335.1 2783 Missense Mutation AAA,AAC K890N NP_001076804.1
NM_013380.3 2783 Missense Mutation AAA,AAC K884N NP_037512.3

View Full Product Details