Product Details

SNP ID
rs144521764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38815670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGTTCTCAGTCGTGGCCTGGAC[C/T]GACTTCACGAGGTCTTGGGTCTGCA
Phenotype
MIM: 600696 MIM: 602518
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ECH1 PubMed Links

Gene Details

Gene
ECH1
Gene Name
enoyl-CoA hydratase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001398.2 1015 Silent Mutation TCA,TCG S310S NP_001389.2
XM_017026448.1 1015 Silent Mutation TCA,TCG S230S XP_016881937.1
Gene
LGALS4
Gene Name
galectin 4
There are no transcripts associated with this gene.

View Full Product Details