Product Details

SNP ID
rs144583939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57925853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTCAATTTAGGCAGATGGCTCC[A/G]TCATAAGGCATCTCCTCCAGTGTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF417 PubMed Links

Gene Details

Gene
ZNF417
Gene Name
zinc finger protein 417
There are no transcripts associated with this gene.

Gene
ZNF418
Gene Name
zinc finger protein 418
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317027.1 2634 UTR 3 NP_001303956.1
NM_001317028.1 2634 UTR 3 NP_001303957.1
NM_001317029.1 2634 UTR 3 NP_001303958.1
NM_001317030.1 2634 UTR 3 NP_001303959.1
NM_133460.2 2634 UTR 3 NP_597717.1
XM_017026304.1 2634 UTR 3 XP_016881793.1
XM_017026305.1 2634 UTR 3 XP_016881794.1
XM_017026306.1 2634 UTR 3 XP_016881795.1
XM_017026307.1 2634 UTR 3 XP_016881796.1
XM_017026308.1 2634 UTR 3 XP_016881797.1
XM_017026309.1 2634 UTR 3 XP_016881798.1
XM_017026310.1 2634 UTR 3 XP_016881799.1
XM_017026311.1 2634 UTR 3 XP_016881800.1
XM_017026312.1 2634 UTR 3 XP_016881801.1
XM_017026313.1 2634 UTR 3 XP_016881802.1
XM_017026314.1 2634 UTR 3 XP_016881803.1

View Full Product Details