Product Details

SNP ID
rs144598692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4048190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGGCGGCGCCGCACTGCTGGCA[C/T]AGGTACGGCTTCTCGCCCGTGTGCT
Phenotype
MIM: 605989 MIM: 605878
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIAS4 PubMed Links

Gene Details

Gene
PIAS4
Gene Name
protein inhibitor of activated STAT 4
There are no transcripts associated with this gene.

Gene
ZBTB7A
Gene Name
zinc finger and BTB domain containing 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317990.1 2518 Silent Mutation CTA,CTG L439L NP_001304919.1
NM_015898.3 2518 Silent Mutation CTA,CTG L439L NP_056982.1
XM_005259570.4 2518 Silent Mutation CTA,CTG L489L XP_005259627.3
XM_005259571.4 2518 Silent Mutation CTA,CTG L439L XP_005259628.1

View Full Product Details