Product Details

SNP ID
rs144608866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19120031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGGTAGGCCAGGACGCCACGGA[A/G]GAAGAGGGGAGTAAGCAGGCCACCC
Phenotype
MIM: 610823
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A42 PubMed Links

Gene Details

Gene
SLC25A42
Gene Name
solute carrier family 25 member 42
There are no transcripts associated with this gene.

Gene
TMEM161A
Gene Name
transmembrane protein 161A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256766.1 1308 Missense Mutation CTC,TTC L344F NP_001243695.1
NM_017814.2 1308 Missense Mutation CTC,TTC L447F NP_060284.1
XM_011528117.2 1308 Missense Mutation CTC,TTC L245F XP_011526419.1
XM_017026916.1 1308 Missense Mutation CTC,TTC L422F XP_016882405.1

View Full Product Details