Product Details

SNP ID
rs144664411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49908290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTGCTCCTTGCGCCGGCCCTC[A/G]CACACCTTGGTCACCTCCTCCACCT
Phenotype
MIM: 609742 MIM: 605815
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL4I1 PubMed Links

Gene Details

Gene
IL4I1
Gene Name
interleukin 4 induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258017.1 1721 Intron NP_001244946.1
NM_001258018.1 1721 Intron NP_001244947.1
NM_152899.1 1721 Intron NP_690863.1
NM_172374.2 1721 Intron NP_758962.1
Gene
NUP62
Gene Name
nucleoporin 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193357.1 1721 Silent Mutation TGC,TGT C506C NP_001180286.1
NM_012346.4 1721 Silent Mutation TGC,TGT C506C NP_036478.2
NM_016553.4 1721 Silent Mutation TGC,TGT C506C NP_057637.2
NM_153718.3 1721 Silent Mutation TGC,TGT C506C NP_714940.1
NM_153719.3 1721 Silent Mutation TGC,TGT C506C NP_714941.1

View Full Product Details