Product Details

SNP ID
rs145046422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52300417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATTGACTTCTAAAGAGTCATGC[G/T]GTGTGATGAAAAAGCCCAGAAGAGA
Phenotype
MIM: 613910
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF480 PubMed Links

Gene Details

Gene
ZNF480
Gene Name
zinc finger protein 480
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297624.1 481 Missense Mutation CGG,CTG R2L NP_001284553.1
NM_001297625.1 481 UTR 5 NP_001284554.1
NM_144684.3 481 Missense Mutation CGG,CTG R2L NP_653285.2
XM_011526465.2 481 Missense Mutation CGG,CTG R2L XP_011524767.1
Gene
ZNF766
Gene Name
zinc finger protein 766
There are no transcripts associated with this gene.

View Full Product Details