Product Details

SNP ID
rs145099125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55174934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGTTTTTAGCCTCCAGGACGAT[A/G]ACGGTGAGCTTCCCGGCCGTGGGGA
Phenotype
MIM: 614566 MIM: 602510 MIM: 600782
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAAF3 PubMed Links

Gene Details

Gene
DNAAF3
Gene Name
dynein axonemal assembly factor 3
There are no transcripts associated with this gene.

Gene
PTPRH
Gene Name
protein tyrosine phosphatase, receptor type H
There are no transcripts associated with this gene.

Gene
SYT5
Gene Name
synaptotagmin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297774.1 1141 Silent Mutation GTC,GTT V254V NP_001284703.1
NM_003180.2 1141 Silent Mutation GTC,GTT V258V NP_003171.2
XM_006723339.3 1141 Silent Mutation GTC,GTT V258V XP_006723402.1
XM_006723340.3 1141 Silent Mutation GTC,GTT V257V XP_006723403.1
XM_006723341.3 1141 Silent Mutation GTC,GTT V255V XP_006723404.1
XM_017027175.1 1141 Silent Mutation GTC,GTT V258V XP_016882664.1
XM_017027176.1 1141 Silent Mutation GTC,GTT V258V XP_016882665.1

View Full Product Details