Product Details
- SNP ID
-
rs145117632
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:36142330 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCTGCGCAGTACAACCCGGAGCC[C/G]CCGGTAAGCCCCCTCTGCAACCAGA
- Phenotype
-
MIM: 114170
MIM: 123995
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CAPNS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2075475] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAPNS1
- Gene Name
- calpain small subunit 1
- Gene
- COX7A1
- Gene Name
- cytochrome c oxidase subunit 7A1
There are no transcripts associated with this gene.
View Full Product Details