Product Details
- SNP ID
-
rs145132549
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:58433481 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTTTCCACACTGGATGCACTCAT[A/G]GGGCCTTTCTTGTGTGTGAACTCTC
- Phenotype
-
MIM: 604074
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF132
PubMed Links
Gene Details
- Gene
- ZNF132
- Gene Name
- zinc finger protein 132
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003433.3 |
2364 |
Missense Mutation |
CAT,TAT |
H655Y |
NP_003424.3 |
View Full Product Details