Product Details

SNP ID
rs145210985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51371971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCAAGAGGATAGCTGAGACCCA[G/T]CCCAGGTAGAAGGACCAGGAGAAGA
Phenotype
MIM: 130410 MIM: 154045 MIM: 606008
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLDND2 PubMed Links

Gene Details

Gene
CLDND2
Gene Name
claudin domain containing 2
There are no transcripts associated with this gene.

Gene
ETFB
Gene Name
electron transfer flavoprotein beta subunit
There are no transcripts associated with this gene.

Gene
LIM2
Gene Name
lens intrinsic membrane protein 2
There are no transcripts associated with this gene.

Gene
NKG7
Gene Name
natural killer cell granule protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005601.3 414 Silent Mutation GGA,GGC G136G NP_005592.1
XM_005258955.3 414 Silent Mutation GGA,GGC G101G XP_005259012.1
XM_006723228.3 414 Silent Mutation GGA,GGC G87G XP_006723291.1

View Full Product Details