Product Details

SNP ID
rs145251123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49514444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATTCTCCTGCCCGCACCGCCTG[C/T]GGGAGCACCTGGAGAGGGGCCGCGG
Phenotype
MIM: 601437
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCGRT PubMed Links

Gene Details

Gene
FCGRT
Gene Name
Fc fragment of IgG receptor and transporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136019.2 781 Missense Mutation CGG,TGG R187W NP_001129491.1
NM_004107.4 781 Missense Mutation CGG,TGG R187W NP_004098.1
XM_005258657.1 781 Missense Mutation CGG,TGG R47W XP_005258714.1
XM_011526627.2 781 Intron XP_011524929.1

View Full Product Details