Product Details

SNP ID
rs145317573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38620406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGGCCAAGGAAAATGCCTATGA[C/T]CTGGAAGCCAACCTGGCTGTCCTGA
Phenotype
MIM: 609596 MIM: 601983
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF3K PubMed Links

Gene Details

Gene
EIF3K
Gene Name
eukaryotic translation initiation factor 3 subunit K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300992.1 Intron NP_001287921.1
NM_001308393.1 Intron NP_001295322.1
NM_013234.3 Intron NP_037366.1
Gene
MAP4K1
Gene Name
mitogen-activated protein kinase kinase kinase kinase 1
There are no transcripts associated with this gene.

View Full Product Details