Product Details

SNP ID
rs145341903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44944846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGGCAGGATGAAGGAGCTGCT[G/T]GAGACAGTGGTGAACAGGACCAGAG
Phenotype
MIM: 608083 MIM: 600745 MIM: 604783
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
APOC2 PubMed Links

Gene Details

Gene
APOC2
Gene Name
apolipoprotein C2
There are no transcripts associated with this gene.

Gene
APOC4
Gene Name
apolipoprotein C4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001646.2 214 Silent Mutation CTG,CTT L58L NP_001637.1
Gene
APOC4-APOC2
Gene Name
APOC4-APOC2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
CLPTM1
Gene Name
CLPTM1, transmembrane protein
There are no transcripts associated with this gene.

View Full Product Details