Product Details

SNP ID
rs145401547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34759297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACAAACATAGGGCTTCTCTCCAG[C/T]GTGAATCCTCATGTGTCGAGTGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF599 PubMed Links

Gene Details

Gene
ZNF599
Gene Name
zinc finger protein 599
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007248.2 1892 Missense Mutation ACT,GCT T502A NP_001007249.1
XM_017026369.1 1892 Missense Mutation ACT,GCT T465A XP_016881858.1

View Full Product Details