Product Details

SNP ID
rs145408161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11454576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAGGAGACCTGCAGCACGCGCT[C/T]GCCCAGGCGATAGCCGTTCAGGCTG
Phenotype
MIM: 603458 MIM: 177060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELAVL3 PubMed Links

Gene Details

Gene
ELAVL3
Gene Name
ELAV like neuron-specific RNA binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001420.3 1437 Missense Mutation AAG,GAG K352E NP_001411.2
NM_032281.2 1437 Missense Mutation AAG,GAG K345E NP_115657.2
XM_011527778.2 1437 Missense Mutation AAG,GAG K351E XP_011526080.1
Gene
PRKCSH
Gene Name
protein kinase C substrate 80K-H
There are no transcripts associated with this gene.

View Full Product Details